Please login to your online account to display your discounted pricing
Fisherbrand™ Polypropylene Desiccator Plates
Withstand heavy use.
Manufacturer: fisherbrand™  EMD256025
 View more versions of this product
Catalog No. S33522A
  • / Each
  • / Case of 10 Each
Description & Specifications
Specifications
For Use With (Application) Unbreakable, economical desiccator plates
Diameter (English) Outer 5.51 in.
Diameter (Metric) Outer 14cm
Holds 24
Product Type Desiccator Plate
Material Polypropylene