missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Polymethylpentene Graduated Cylinders

Calibrated “To Contain/To Deliver” at 20°C

Manufacturer:  Fisherbrand™ N3004500100

 View more versions of this product

Catalog No. 03-007-33



 

Description

Description

  • Transparent cylinders with large molded-in graduations
  • Easy to read
  • Large rounded bases ribbed for reinforcement and stability
  • Generous pour spout; 10mL size has a conical top for easier filling and pouring
  • Non-wetting
  • Chemical, heat and impact resistant
  • Window-clear

Specifications

Specifications

100mL
Circular, Ribbed
1mL
1mL
PMP (Polymethylpentene)
To contain/to deliver
Yes
Documents