missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisherbrand™ Autoclavable HD Waste Bags
Biohazard symbol printed bags to ensure safe handling and disposal of biohazardous waste.
Manufacturer: Fisherbrand™ 01831B
Description
Biohazard symbol printed bags to ensure safe handling and disposal of biohazardous waste.
- All bags supplied with autoclavable elastic steam-venting closures except 1.5mil-thick plain clear bags
- Polyethylene
- Temerature range 260degrees F
- Available in 4 colors/styles
Specifications
48 x 38 in. (122 x 97 cm) | |
127°C | |
Yes | |
Elastic steam venting | |
Printed in English and Spanish | |
122 cm | |
38 in. |
260°F | |
4mil | |
OSHA, 29 CFR1910.1030 | |
Printed Red | |
48 in. | |
PE (Polyethylene) | |
97 cm |