Please login to your online account to display your discounted pricing

Fisherbrand™ Poly Boots

Great for splash protection

$28.70 - $49.67

Specifications

Color Clear
Material Polypropylene
For Use With (Application) Ideal for splash protection and blood spills or bodily fluid cleanup
Style Calf-high
View More Specs
Products
Catalog Number Mfr. No. Weight (English) Weight (Metric) Product Type Size Price Quantity    

02-740A

 
aep industries inc
2BT1MF
19 lb. 8.61kg Heavyweight Boot Covers Medium Pack of 100 Each for $28.70
Case of 10 Pack for $265.90

02-740D

 
aep industries inc
2BT3LF
43 lb. 19.5kg Extra Heavyweight Boot Covers Large Pack of 100 Each for $49.67
Case of 10 Pack for $460.20
Description & Specifications

Specifications

Color Clear
Material Polypropylene
For Use With (Application) Ideal for splash protection and blood spills or bodily fluid cleanup
Style Calf-high

Ideal for blood spills or bodily fluid cleanup.

  • Constucted of clear polypropylene
  • Calf-high boots protect shoes