Fisherbrand™ Pocket Closed Metal Case Liquid-In-Glass Thermometers
Manufacturer:  Hb Instrument B607701620
 View more versions of this product
Catalog No. 13201696
Specifications Description & Specifications
Specifications
1°C
Field studies, waste water treatment and any "on-the-go" applications
Multilanguage instructions, statement of accuracy
160mm
1
-10° to +110°C
ISO/IEC 17025:2005, A2LA, NIST
Total
6.3 in.
White Back Glass
Not Applicable
Blue Organic