missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Pocket Closed Metal Case Liquid-In-Glass Thermometers

Manufacturer:  Fisher Scientific B607701920

 View more versions of this product

Catalog No. 13-202-505


 

Specifications Description & Specifications

Specifications

2°F
Field studies, waste water treatment and any "on-the-go" applications
Multilanguage instructions, statement of accuracy
160mm
2
Not Applicable
ISO/IEC 17025:2005, A2LA, NIST
Total
6.3 in.
White Back Glass
-30° to +120°F
Blue Organic