Fisherbrand™ Plastic pH Strips
Laboratory grade plastic pH strips in durable and easy to open flip-top vials
Manufacturer:  Fisherbrand™ B60-SHTRG-000060-VPS
 View more versions of this product
Catalog No. 13-640-520
Description
Description
Laboratory grade plastic pH strips in durable and easy to open flip-top vials
  • Easy-to-use — simply dip strip in solution to be tested and match to color chart immediately
  • Single color match with full 0.5 in. (1.27cm) matching area for quick and accurate matches
  • Flip-top vial stays closed when dropped, but is easy to open with one-hand
  • Sturdy vial keeps strips safe from the elements
  • Full pH range (0.0-14.0) is available in a universal pH strip and finer measurements across the pH range
  • 100 strips per vial; pack of 6 vials
Order Info
Box 6 Vials, 100 strips per vial.
Specifications
Specifications
For quick and accurate matches
100 strips per vial, 6 vials per pack
Room Temperature
0 to 6
0.5
0 to 6
pH Strip
Documents