Please login to your online account to display your discounted pricing

Fisherbrand™ Plain Glass Microslides


Individually selected for consistent quality

Manufacturer: fisherbrand™  ER500-600621T

 View more versions of this product

Catalog No. 12-550D

  • / Gross of 144 Each
  • / Case of 10 Gross
 

Description & Specifications

Specifications

Edges Ground Edges
Length (Metric) 75mm
Thickness 1mm
Coating Chemical Resistant
Material Glass
Width (Metric) 25mm
Color White