Please login to your online account to display your discounted pricing

Fisherbrand™ Pipet Tips


General purpose for universal applications

Manufacturer: fisherbrand™  111FISEMD

 View more versions of this product

Catalog No. S63213

  • / Pack of 1000 Each
  • / Case of 10 Pack
 

Description & Specifications

Specifications

Product Type Pipet tips
Material Polypropylene
Length (English) 2.75 in.
Length (Metric) 6.9cm
For Use With (Equipment) Research grade pipetters
Sterility Nonsterile
Autoclavable Yes
Tip Style Redi-Tip™ Tip
Packaging Bulk
Certifications/Compliance RNase, DNase, DNA, pyrogen-free
Volume (Metric) 101 to 1000μL
Color Blue