Please login to your online account to display your discounted pricing

Fisherbrand™ Pigskin Palm Gloves

Repel moisture and do not stiffen even after soaked in water

$73.32 - $73.32

Specifications

Size Large
For Use With (Application) pipe, brick and other material handling, construction, farming and general maintenance
Glove Cuff Style Shirred Elastic
Hem Color Brown
Length (English) Cuff 2.5 in.
View More Specs
Products
Catalog Number Mfr. No. Material Price Quantity    

19-013508

 
superior glove works ltd
76PB
Premium Grade Pig Skin Leather Dozen of 12 Pair for $73.32
Description & Specifications

Specifications

Size Large
For Use With (Application) pipe, brick and other material handling, construction, farming and general maintenance
Glove Cuff Style Shirred Elastic
Hem Color Brown
Length (English) Cuff 2.5 in.
Length (Metric) Cuff 6cm
Liner Material Cotton

Highly versatile pigskin leather palm work gloves are suited for construction, machine operation, utility work, and many general applications.

  • Cotton lined with traditional striped fabric back
  • Durable and thin, providing greater dexterity
  • Gunn-cut style with wing thumb, shirred elastic back, and leather pull patch
  • Leather covers index finger, thumb, knuckle, and fingertips
  • With brown cuff hem
  • Available in two grades of leather with a 2.5 in. (6cm) rubberized safety cuff
  • Men's size