missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisherbrand™ PFA Safety Coated Liquid-In-Glass Partial Immersion Thermometers
In the event of breakage, the PFA fluoropolymer safety coating keeps the liquid and glass encapsulated.
Supplier: Fisherbrand™ B607100220
Description
- Contain blue organic liquid against lead-free white back glass
- Traceable to NIST, individually serialized
- Include multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy and Traceability
- Tested and inspected by our USA quality team
- Coated thermometers are just as accurate as uncoated thermometers, but are not recommended for timed tests as the coating can slow reaction time
- Ideal for use in chemical laboratories, universities, scientific institutions, food and beverage processing, petrochemical laboratories, pharmaceutical industry, medical industry, manufacturers, dairies, breweries, and distilleries
Specifications
ISO/IEC 17025:2005, A2LA, NIST | |
Partial Immersion | |
200 mm | |
Organic | |
-10°C | |
-10°C to +110°C | |
1°C |
Chemical laboratories, universities, scientific institutions, food and beverage processing, petrochemical laboratories, pharmaceutical industry, medical industry, manufacturers, dairies, breweries and distilleries | |
7.9 in. | |
Glass | |
110°C | |
— | |
±1.5°C | |
50 mm |