Please login to your online account to display your discounted pricing
Fisherbrand™ Permanent Markers
Write on all surfaces
Manufacturer: fisherbrand™  CON3050
 View more versions of this product
Catalog No. 13-383
  • / Pack of 12 Each
Description & Specifications
Specifications
Product Type Permanent Marker
Width (Metric) Line 0.3mm
Color Assorted
For Use With (Application) Perfect marking pens for lab samples, microscope slides, photographic slides, glass and plastic beakers, and plastic sample bags
Includes Six opaque black, two vivid red, two blue, and two green pens with seal cap
Material Stainless Steel
Quantity 12 Pack
Fisherbrand™ Permanent Marking Pens write on glass, film, wood, plastic, metal, rubber, and porcelain. Perfect for marking sample bags, microscope or photographic slides, laboratory glassware and more. Write on all surfaces.
  • Produce precise, ultrathin lines 0.3mm wide
  • Waterproof and fadeproof, permanent ink dries almost instantly
  • An airtight ring seal cap prevents dry-out and ensures instant writing even after lengthy storage
  • Unique stainless-steel long leg allows precise writing in a confined space