Please login to your online account to display your discounted pricing
Fisherbrand™ Black Phenolic PolyCone Cap
Form an exceptionally tight seal and offer a good chemical barrier. Fisherbrand™ Black Phenolic PolyCone Caps are used from liquids, to soaps, to macro-invertebrates.
Manufacturer: fisherbrand™  263831
 View more versions of this product
Catalog No. 02-911-727
  • / Case of 1600 Each
Description & Specifications
Specifications
Product Type PE Cone Lined Cap
Closure Color Black
Closure Material Phenolic
Closure Size 38/400
Closure Type Screw
Material Phenolic
Liner Material PolyCone
Quantity 1600/Cs.
Closure Liner Polyseal
Neck Finish 38-400
  • Liner molds itself around the sealing areas of finish as cap is screwed on
  • Eliminates leakage, evaporation, contamination, binding and backing off