Please login to your online account to display your discounted pricing

Fisherbrand™ Black Phenolic PolyCone Caps


Forms an exceptionally tight seal and offer a good chemical barrier. Fisherbrand™ Black Phenolic PolyCone Caps are crack resistant.

Manufacturer: fisherbrand™  263828L

 View more versions of this product

Catalog No. 02-912-061

  • / Pack of 12 Each
  • / Case of 48 Pack
 

Description & Specifications

Specifications

Certifications/Compliance ISO 9001
Closure Material Phenolic
Closure Liner Polyseal
Color Black
Description Cap
Material Phenolic
No. per Case 48 Cs.
Packaging Packed in 48 bags of 12
Quantity 576EA/Cs.
Type Cap, PE Cone Lined
For Use With (Equipment) Bottle
Closure Color Black
Liner Material PolyCone
Neck Finish 28-400
Shape Round
Closure Type Screw

  • Liner molds itself around sealing areas of the finish as the cap is screwed on, virtually eliminating leakage, evaporation, contamination, binding and backing off