Please login to your online account to display your discounted pricing

Fisherbrand™ Black Phenolic PolyCone Caps


Forms an exceptionally tight seal and offer a good chemical barrier. Fisherbrand™ Black Phenolic PolyCone Caps are crack resistant.

Manufacturer: fisherbrand™  263824L

 View more versions of this product

Catalog No. 02-912-058

  • / Case of 576 Each
 

Description & Specifications

Specifications

Closure Color Black
Type PE Cone Lined Caps
Packaging Packed in 48 bags of 12
For Use With (Equipment) Bottle
Closure Type Screw
Liner Material PolyCone
Shape Round
Neck Finish 15-425
Material Phenolic
Closure Material Phenolic
Quantity 576EA/Cs.
Color Black
Certifications/Compliance ISO 9001
Closure Liner Polyseal
No. per Case 576 Cs.

  • Liner molds itself around sealing areas of the finish as the cap is screwed on, virtually eliminating leakage, evaporation, contamination, binding and backing off