missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Black Phenolic PolyCone Cap

Forms an exceptionally tight seal and offers a good chemical barrier. Fisherbrand™ Black Phenolic PolyCone Caps are crack resistant.

$10.73 - $10.83

Specifications

Certifications/Compliance ISO 9001
Closure Liner Polyseal
Color Black
Material Phenolic
For Use With (Equipment) Bottle
View More Specs

Products 2
Catalog Number Mfr. No. Closure Size Price Quantity  
Catalog Number Mfr. No. Closure Size Price Quantity  
02-912-059 View Documents Fisherbrand™
263825L
Pack of 12 Each for $10.83
Case of 48 Pack for $519.84
 
02-912-061 View Documents Fisherbrand™
263828L
Pack of 12 Each for $10.73
Case of 48 Pack for $515.04
 
Description

Description

PolyCone Lined Phenolic Caps form an exceptionally tight seal and offer a good chemical barrier. This liner molds itself around the sealing areas of the finish as the cap is screwed on, virtually eliminating leakage, evaporation, contamination, binding and backing off. Great for everything from liquids, to soaps, to macro-invertebrates.

  • Liner molds itself around sealing areas of the finish as the cap is screwed on, virtually eliminating leakage, evaporation, contamination, binding and backing off
  • Great for everything from liquids, to soaps, to macro-invertebrates

Specifications

Specifications

ISO 9001
Black
Bottle
Polyseal
Phenolic
PE Cone Lined Cap,PolyCone Lined Cap