Fisherbrand™ Black Phenolic PolyCone Cap
Form an exceptionally tight seal and offer a good chemical barrier. Fisherbrand™ Black Phenolic PolyCone Caps are used from liquids, to soaps, to macro-invertebrates.
Manufacturer:  Fisherbrand™ 263826
 View more versions of this product
Catalog No. 02-911-803
Specifications
Description
Cap, Polyethylene; Fisherbrand; LDPE; PE cone Black; Stress-crack resistant; General-purpose; 20mm-400 screw cap size; 5500/cs.
  • Liner molds itself around the sealing areas of finish as cap is screwed on
  • Eliminates leakage, evaporation, contamination, binding and backing off
Description & Specifications
Specifications
Phenolic
Phenolic
Cap, PE Cone Lined
20/400
20-400
Polyseal
5500/Cs.
Black
PolyCone
Screw