Fisherbrand™ Ostwald-Folin Pipets
High-volume accuracy at a reasonable price.
Description
High-volume accuracy at a reasonable price.
- Tapered bulbs
- Borosilicate glass with permanent amber markings
- Tuf-Temp™-treated to reduce chipping
- Accuracy printed on each pipet
- Accuracy equivalent to that of Class A volumetric
Order Info
Available as PersonaPet™ personalized pipets
Compliance
Calibrated “To Deliver/Blow Out.” Meet Fed. Spec. NNN-P-340B (or latest revision).
Specifications
±0.006 mL | |
NNN-P-340B | |
3 sec. | |
0.3 mL |
Yes | |
Borosilicate Glass | |
0.0101 oz. | |
Ostwald Folin Pipet |