Fisherbrand™ Ostwald-Folin Pipets
High-volume accuracy at a reasonable price
Manufacturer:  Fisherbrand™ 1138232C
 View more versions of this product
Catalog No. 11-382-32C
Specifications Description & Specifications
Specifications
±0.006mL
0.0084 oz., 0.25mL
3 sec.
0.25mL
Ostwald Folin Pipet
Yes
NNN-P-340B
0.0084 oz.
Borosilicate Glass