Fisherbrand™ Opaque Sterile Sampling Bag
Ideal for photosensitive sampling applications
Manufacturer: Fisherbrand™ EPN4590
Description
Black-pigmented resin provides a total light barrier that protects photosensitive substances such as vitamins, chemicals, pharmaceuticals, food supplements, plant extracts, and biological tissues. Ideal for processing abrasive chemicals and long-term storage.
- Made from highly resistant, LDPE virgin polyethylene tubing
- RNase, DNase, and pyrogen free
- LDPE tubing extruded at temperatures from 220° to 240°C, guaranteeing internal sterility
- Product sterility verified through random testing with independent labs
- With write-on strip and safety-tab closures
- Thickness, 3mil
Certifications
Sterility certificate available on request
Compliance
Complies with EPA, FDA, USDA, and HACCP requirements
Specifications
9 x 4.5 in. (229 x 114mm) | |
3 mil | |
9 in. | |
LDPE (Low-density Polyethylene) | |
114mm |
220° to 240°C | |
18 oz. | |
229mm | |
4.5 in. |