Fisherbrand™ Octagon Magnetic Stir Bars
Octagon magnetic stirring bars with integral pivot ring are the most commonly used shape
$7.09 - $13.84
Specifications
Case Quantity 50
Coating PTFE
Material Alnico Magnet
Product Type Stir Bar
Shape Octagon
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Diameter (English) Diameter (Metric) Length (English) Length (Metric) Price Quantity  
Catalog Number Mfr. No. Diameter (English) Diameter (Metric) Length (English) Length (Metric) Price Quantity  
S68071 Fisherbrand™
F371105013/EMD
0.35 in. 9mm 2 in. 50.8mm Each for $13.84
S68069 Fisherbrand™
F371105001/EMD
0.31 in. 8mm 1 in. 25.4mm Each for $7.09
S68070 Fisherbrand™
F371105006/EMD
0.31 in. 8mm 1.5 in. 38mm Each for $7.83
Specifications
Description
  • Alnico magnet encapsulated in FDA Grade, USP Class VI Teflon™ PTFE low friction coating; durable and inert
  • Interrupted profile provides greater surface area and added turbulence when compared to the smooth surface of cylindrical bars
  • Pivot ring aids in reducing friction and chattering (F37110-1218 does not have a pivot ring)
  • Autoclavable
  • Manufactured in a registered ISO 9001:2008, as verified by SGS certification
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Description & Specifications
Specifications
50
Alnico Magnet
Octagon
PTFE
Stir Bar