Fisherbrand™ Nonsterile Plastic Culture Tubes
Extra-heavy wall construction is ideal for numerous testing applications
Manufacturer:  Fisherbrand™ 1496110A
 View more versions of this product
Catalog No. 14-961-10A
Description
Description
Extra-heavy wall construction is ideal for numerous testing applications
  • Round bottom
  • Special molding avoids leaks and cracks
  • 12 O.D. × 75mm long tube has 5mL capacity and heavier wall than competitive tubes
  • Slight lip at top provides seal when used with Cat. No. 14-961-12 Snap Cap (sold separately)
Order Info
Select 12 × 75mm tubes available with special one-directional, oriented-box packaging (see ordering information). Caps for 12mm and 17mm tubes available separately (14-961-12, 14-961-15).
Specifications
Specifications
Clear
Yes
1000/Cs.
5mL
75mm
Culture Test Tube
Non-sterile
12mm
Ideal for numerous testing applications
5mL
Without Cap
Polystyrene
Round Bottom
Documents
Accessories