Fisherbrand™ Nonsterile Plastic Culture Tubes
Extra-heavy wall construction is ideal for numerous testing applications
Manufacturer:  Fisherbrand™ 1496120
 View more versions of this product
Catalog No. 14-961-20
Description
Description
Extra-heavy wall construction is ideal for numerous testing applications
Order Info
Select 12 × 75mm tubes available with special one-directional, oriented-box packaging (see ordering information). Caps for 12mm and 17mm tubes available separately (14-961-12, 14-961-15).
Specifications
Specifications
Clear
Yes
Snap cap
5mL
75mm
Culture Test Tube
Non-sterile
12mm
Ideal for numerous testing applications
5mL
Snap
Polystyrene
Round Bottom
Documents