Please login to your online account to display your discounted pricing
Fisherbrand™ Nonsterile Threaded End Microcentrifuge Tubes without Caps
Choose skirted- or conical-bottom style
Manufacturer: fisherbrand™  519GRDFIS
 View more versions of this product
Catalog No. 02-681-343
  • / Pack of 500 Each
  • / Case of 10 Pack
Description & Specifications
Specifications
Closure Type Screw
Capacity (Metric) 2mL
Material Plastic
Color Natural
Sterile No
Shape Skirted
Max. Force 18000xG
Accessories
Accessories for Catalog Number: 02681343
Item Mfr. No. Item Description Price Per Unit Quantity  
05-407-22
Fisherbrand™ Microcentrifuge Tube Accessory Micro Tube Tool
fisherbrand™
BIP4000
For use in capping or decapping Fisherbrand Microcentrifuge Tubes with Snap Caps Pack of 4 Each for $14.20
Case of 25 Pack for $355.00