Please login to your online account to display your discounted pricing
Fisherbrand™ Nonsterile Threaded End Microcentrifuge Tubes without Caps
Choose skirted- or conical-bottom style
Manufacturer: fisherbrand™  528FIS
 View more versions of this product
Catalog No. 02-681-338
  • / Pack of 500 Each
  • / Case of 10 Pack
Description & Specifications
Specifications
Item Description Without caps
Capacity (Metric) 1.5mL
Color Natural
Material Plastic
Max. Force 18000xG
Sterile No
Shape Skirted
Closure Type Screw
Accessories
Accessories for Catalog Number: 02681338
Item Mfr. No. Item Description Price Per Unit Quantity