Fisherbrand™ Nonmercury Verification Thermometers
To verify sample temperatures in freezers, refrigerators, and incubators with traceability to NIST.
Supplier: Fisherbrand™ B610030520
Description
Colored organic fill against white back; PFA safety coated.
- 30mL bottle size, choice of bottle fills
- Individual serial numbers and a Statement of Accuracy traceable to NIST
- Tested in a triple accredited/registered ISO/IEC 17025:2005, A2LA laboratory
Applications:
Ideal for the chemical, petrochemistry, pharmaceutical, medical, and cosmetic industries; producers of chemicals, universities, dairies, breweries, distillers, beer cellars, refrigeration houses, hot houses, scientific institutes, blood banks, refrigeration/freezer/incubator equipment producers, and producers of laboratory equipment
Specifications
ISO/IEC 17025:2005, A2LA, NIST | |
145 mm | |
Verification Thermometer: Oven | |
20°C | |
±1.5°C and ±2°C above 100°C |
Ideal for the chemical, petrochemistry, pharmaceutical, medical, and cosmetic industries; producers of chemicals, universities, dairies, breweries, distillers, beer cellars, refrigeration houses, hot houses, scientific institutes, blood banks, refrigeration/freezer/incubator equipment producers, and producers of laboratory equipment. | |
Glass | |
130°C | |
20°C to 130°C |