Please login to your online account to display your discounted pricing

Fisherbrand™ Natural Color Micro Pipet Tips

For use with micro volume pipetters

$71.00 - $126.90

Specifications

Color Natural
For Use With (Equipment) Microvolume pipetter
Material Polypropylene
Product Type Micro Tip
Product Type Micro Tip
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Sterility Volume (Metric) Price Quantity    

S66804

 
fisherbrand™
H-104-96R-F
Hinged Rack 960 Pack Nonsterile 0.1 to 10μL Pack of 960 Each for $126.90
Case of 5 Pack for $634.55

S66803

 
fisherbrand™
104-FIS/EMD
Bulk 1000 Pack Nonsterile 0.1 to 10μL Pack of 1000 Each for $71.00
Case of 10 Pack for $710.00
Description & Specifications

Specifications

Color Natural
For Use With (Equipment) Microvolume pipetter
Material Polypropylene
Product Type Micro Tip
Product Type Micro Tip

  • Natural color
  • S66803 is packaged 1000 tips per pack; others have 960 tips

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.