Please login to your online account to display your discounted pricing

Fisherbrand™ Natural Color Micro Pipet Tips


For use with micro volume pipetters

Manufacturer: fisherbrand™  104-FIS/EMD

 View more versions of this product

Catalog No. S66803

  • / Pack of 1000 Each
  • / Case of 10 Pack
 

Description & Specifications

Specifications

Product Type Micro Tip
Sterility Nonsterile
For Use With (Equipment) Microvolume pipetter
Material Polypropylene
Volume (Metric) 0.1 to 10μL
Color Natural
Tip Style Micro Point
Quantity 1000 Pack
Packaging Bulk