Please login to your online account to display your discounted pricing

Fisherbrand™ Narrow Mouth Boston Rounds with Screw-Caps

Machine-made flint glass bottles.

Manufacturer: fisherbrand™  W014046/EMD

 View all options for this product

Catalog No. S31665F


Description & Specifications


Capacity 32 oz., 1000mL
Capacity (Metric) 1000mL
Capacity (English) 32 oz.
Closure Screw
Closure Material Plastic
Closure Liner Poly/Vinyl
Color Amber
Material Flint Glass

  • Clear or amber
  • Include plastic screw caps with polyvinyl liners