missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Narrow Mouth Boston Rounds with Screw-Caps

Machine-made flint glass bottles.

$29.05 - $162.00

Specifications

Closure Liner Poly/Vinyl
Closure Material Plastic
Capacity (English) 4 oz.
Capacity (Metric) 125mL
Closure Type Screw
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Capacity Color Price Quantity  
Catalog Number Mfr. No. Capacity Color Price Quantity  
S31840F Fisherbrand™
W014022/EMD
4 oz., 125mL Clear Case of 24 Each for $68.85
S31841F Fisherbrand™
W014024/EMD
8oz., 236mL Clear Case of 12 Each for $162.00
S31842F Fisherbrand™
W014026/EMD
16 oz., 500mL Clear Case of 12 Each for $29.05
S31843F Fisherbrand™
W014029/EMD
32 oz., 1000mL Clear Case of 12 Each for $67.50
Specifications

Description

  • Clear or amber
  • Include plastic screw caps with polyvinyl liners

Description & Specifications

Specifications

Poly/Vinyl
4 oz.
Screw
Plastic
125mL
Flint Glass