missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Multistage Cylinder Regulators

For noncorrosive gases

Manufacturer:  Genstar Technologies Co 10575136

 View more versions of this product

Catalog No. 10-575-136


 

Alert:

Not for use with corrosive, toxic or high-purity gases.


Specifications Description & Specifications

Specifications

Brass
1 to 125psi
Noncorrosive gases, requiring maintenance of a constant delivery pressure until cylinder contents are empty
Dual-scale
580
3000psi
Nylon Fabric, Nylon
Multistage
Outlet needle valve with 0.25 in. NPTM thread
Neoprene
Argon, Helium, Nitrogen
O2 and CO2 (with CO2 <7%)
5 to 200psi
Yes
Orthane™
Cylinder regulator
Accessories