Please login to your online account to display your discounted pricing

Fisherbrand™ Multistage Cylinder Regulators


For noncorrosive gases

Manufacturer: genstar technologies co  10575149

 View more versions of this product

 Alert

Not for use with corrosive, toxic or high-purity gases.

Catalog No. 10-575-149

  • / Each
 

Description & Specifications

Specifications

Inlet 500
Bonnet Material Brass
Connection Type Outlet needle valve with 0.25 in. NPTM thread
Delivery Pressure 1 to 40psi
Diaphragm Material Neoprene
For Use With (Application) Noncorrosive gases, requiring maintenance of a constant delivery pressure until cylinder contents are empty
Gases Measured Oxygen mix, medical, Oxygen, Carbon Dioxide, (with CO2 < 7%)
Gauge Dual-scale
Includes O2 and CO2 (with CO2 <7%)
Outlet Gauge Pressure 2 to 60psi
Pressure Range (Inlet) 3000psi
Relief Valve Yes
Seal Material Nylon Fabric, Nylon
Seat Material Orthane™
No. of Stages Multistage
Product Type Cylinder regulator