Please login to your online account to display your discounted pricing

Fisherbrand™ Microcentrifuge Tubes with Tether Cap


Reduce sample contamination by keeping cap with tube

Manufacturer: fisherbrand™  TC529FIS

 View more versions of this product

Catalog No. 02-707-358

  • / Pack of 500 Each
  • / Case of 10 Pack
 

Description & Specifications

Specifications

For Use With (Application) Suitable for vapor-phase liquid nitrogen storage, boiling and autoclaving
Capacity (Metric) 1.5mL
Packaging Bag
Material Polypropylene
Color Natural
Shape Skirted
Max. Force 20000xG
Closure Material Polypropylene
Autoclavable Yes
Closure Type Tether Cap
Sterile No
Accessories

Accessories for Catalog Number: 02707358

Item Mfr. No. Item Description Price Per Unit Quantity  
05-407-22
Fisherbrand™ Microcentrifuge Tube Accessory Micro Tube Tool
fisherbrand™
BIP4000
For use in capping or decapping Fisherbrand Microcentrifuge Tubes with Snap Caps Pack of 4 Each for $14.20
Case of 25 Pack for $355.00