Please login to your online account to display your discounted pricing

Fisherbrand™ Microcentrifuge Tubes with Tether Cap


Reduce sample contamination by keeping cap with tube

Manufacturer: fisherbrand™  TC522SFIS

 View more versions of this product

Catalog No. 02-707-355

  • / Pack of 500 Each
  • / Case of 5 Pack
 

Description & Specifications

Specifications

Packaging Bag
Material Polypropylene
Sterile Yes
For Use With (Application) Suitable for vapor-phase liquid nitrogen storage, boiling and autoclaving
Closure Material Polypropylene
Shape Round
Capacity (Metric) 2mL
Autoclavable Yes
Closure Type Tether Cap
Color Natural
Max. Force 20000xG
Accessories

Accessories for Catalog Number: 02707355

Item Mfr. No. Item Description Price Per Unit Quantity  
05-407-22
Fisherbrand™ Microcentrifuge Tube Accessory Micro Tube Tool
fisherbrand™
BIP4000
For use in capping or decapping Fisherbrand Microcentrifuge Tubes with Snap Caps Pack of 4 Each for $14.20
Case of 25 Pack for $355.00