Please login to your online account to display your discounted pricing

Fisherbrand™ Microcentrifuge Tubes with Tether Cap


Reduce sample contamination by keeping cap with tube

Manufacturer: fisherbrand™  TC515SFIS

 View more versions of this product

Catalog No. 02-707-353

  • / Pack of 500 Each
  • / Case of 5 Pack
 

Description & Specifications

Specifications

Closure Material Polypropylene
Sterile Yes
Closure Type Tether Cap
Max. Force 20000xG
Material Polypropylene
Capacity (Metric) 1.5mL
Packaging Bag
Shape Round
For Use With (Application) Suitable for vapor-phase liquid nitrogen storage, boiling and autoclaving
Color Natural
Autoclavable Yes
Accessories

Accessories for Catalog Number: 02707353

Item Mfr. No. Item Description Price Per Unit Quantity  
05-407-22
Fisherbrand™ Microcentrifuge Tube Accessory Micro Tube Tool
fisherbrand™
BIP4000
For use in capping or decapping Fisherbrand Microcentrifuge Tubes with Snap Caps Pack of 4 Each for $14.20
Case of 25 Pack for $355.00