Please login to your online account to display your discounted pricing

Fisherbrand™ Microcentrifuge Tubes with Tether Cap


Reduce sample contamination by keeping cap with tube

Manufacturer: fisherbrand™  TC520GRDSFIS

 View more versions of this product

Catalog No. 02-707-361

  • / Pack of 500 Each
  • / Case of 5 Pack
 

Description & Specifications

Specifications

Capacity (Metric) 2mL
Color Natural
Material Polypropylene
Max. Force 20000xG
Sterile Yes
For Use With (Application) Suitable for vapor-phase liquid nitrogen storage, boiling and autoclaving
Packaging Bag
Shape Skirted
Closure Material Polypropylene
Closure Type Tether Cap
Autoclavable Yes
Accessories

Accessories for Catalog Number: 02707361

Item Mfr. No. Item Description Price Per Unit Quantity