Please login to your online account to display your discounted pricing
Fisherbrand™ Microcentrifuge Foam Tube Racks
Float in water bath with full load of tubes and samples
Manufacturer: fisherbrand™  BIP0020
 View more versions of this product
Catalog No. 05-664-15C
  • / Pack of 6 Each
  • / Case of 6 Pack
Description & Specifications
Specifications
Positions 30.0
Color Gray
Product Type Rack, Microcentrifuge Tube
Autoclavable No
Holds 250 and 400μL Tubes
Autoclavable No
Color Gray
Quantity 6 /pack
Material Foam
Tube Size 250 and 400μL
Positions 30.0