missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Microcentrifuge Foam Tube Racks

Float in water bath with full load of tubes and samples

Manufacturer:  Fisherbrand™ BIP0021

 View more versions of this product

Catalog No. 05-664-15D




Description

Description

  • Made from chemical- and solvent-resistant polyester foam
  • Resilient wells grip tubes securely
  • Flame retardant
  • Capacity: 30 wells, five rows of six sockets per rack

Order Info

Weigh less than 2 oz. (56.7g)

Specifications

Specifications

Not Autoclavable
Tubes (500μL, 1.5mL and 1.8mL)
30
Microcentrifuge Tube Rack
Gray
Foam
6/Pk.
Documents