missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Micro Slide Storage Cabinet Bases

Bases for Fisherbrand Micro Slide Storage Cabinets

Manufacturer:  Fisherbrand™ 07212111

 View more versions of this product

Catalog No. 07-212-111



Description

Description

Bases for Fisherbrand Micro Slide Storage Cabinets

For use with Fisherbrand Micro Slide Storage Cabinets (Cat. No. 07-212-100 through -103)

  • Optional bases in models with legs or without legs
  • Available in dark beige, tan, green, or white
  • 47.6L x 40W x 12.1cm H (18.75 x 15.75 x 4.75 in.)
  • Specifications

    Specifications

    Metal
    18.75 in.
    Fisherbrand Micro Slide Storage Cabinets
    120.65mm
    4.75 in.
    400.05mm
    Tan
    476.25mm
    Microscope slide
    Base
    15.75 in.
    SDS
    Documents