Please login to your online account to display your discounted pricing

Fisherbrand™ Micro Blood Collecting Tubes


With tapered bottom tips to facilitate micro blood sample collection

Manufacturer: fisherbrand™  K42F0266815

 View more versions of this product

Catalog No. 02-668-15

  • / Pack of 100 Each
  • / Case of 6 Pack
 

Description & Specifications

Specifications

Product Type Micro Blood Collecting Tubes
Capacity (Metric) 250μL
Diameter (Metric) Inner 1.44 to 1.76mm
Applications Natelson and Caraway techniques
Additive None
Color Blue
Length (Metric) Tube 147mm
Accessories

Accessories for Catalog Number: 0266815

Item Mfr. No. Item Description Price Per Unit Quantity