Please login to your online account to display your discounted pricing

Fisherbrand™ Micro Blood Collecting Tubes


With tapered bottom tips to facilitate micro blood sample collection

Manufacturer: fisherbrand™  K42E0266810

 View more versions of this product

Catalog No. 02-668-10

  • / Pack of 100 Each
  • / Case of 6 Pack
 

Description & Specifications

Specifications

Product Type Micro Blood Collecting Tubes
Color Red
Applications Natelson and Caraway techniques
Length (Metric) Tube 147mm
Additive Ammonium Heparin
Capacity (Metric) 250μL
Diameter (Metric) Inner 1.44 to 1.76mm
Accessories

Accessories for Catalog Number: 0266810

Item Mfr. No. Item Description Price Per Unit Quantity  
02-678
Fisherbrand™ Hemato-Seal™ Capillary Tube Sealant
fisherbrand™
K4302678
Nondrying, nonhardening, nontoxic compound for sealing hematocrit and other blood collecting tubes Pack of 10 Each for $75.43
Case of 10 Pack for $754.30