Fisherbrand™ Measuring (Mohr) Long-Tip Pipets
Long, tapered delivery tips fit easily into small vials, tubes, or cuvets.
Manufacturer:  Fisherbrand™ 136648J
 View more versions of this product
Catalog No. 13-664-8J
Description
Description
Long, tapered delivery tips fit easily into small vials, tubes, or cuvets.
  • Graduations visible when pipets are in small vials
  • Accuracy printed on each pipet
  • Borosilicate glass
  • Permanent amber markings
  • Tuf-Temp™-treated to reduce chipping
  • Available as PersonaPet™ personalized pipets
Compliance
Meet requirements of Fed. Spec. NNN-P-350C, Style II-Long Taper Tip, except tip lengths are 55 to 60mm. Calibrated “To Deliver” to baseline.
Specifications
Specifications
±0.02mL
0.06 oz., 2mL
White
Yes
Long
2mL
0.06 oz.
NNN-P-350C
White
0.01mL
Measuring Pipets
Borosilicate Glass