Fisherbrand™ Manual Student Burets
With blue markings
Manufacturer:  Fisherbrand™ 703700A
 View more versions of this product
Catalog No. 03-700A
Specifications Description & Specifications
Specifications
For both educational and routine laboratory work
Glass
0.05mL
10mL
0.33 oz., 10mL
0.33 oz.
±0.05mL
Manual Buret
Straight stopcock, blue markings
Accessories