Please login to your online account to display your discounted pricing
Fisherbrand™ Manual Student Burets
With blue markings
$14.90 - $187.80
Specifications
Product Type Manual Buret
For Use With (Application) For both educational and routine laboratory work
Includes Straight stopcock, blue markings
Stopcock Material Glass
View More Specs
Includes:  Straight standard taper glass stopcock.
Products
Catalog Number Mfr. No. Capacity (English) Tolerance (Metric) Capacity (Metric) Subdivision Price Quantity    
03-700A
  Accessories
fisherbrand™
703700A
0.33 oz. ±0.05mL 10mL 0.05mL Each for $148.73
03-700B
  Accessories
fisherbrand™
703700B
0.84 oz. ±0.06mL 25mL 0.06mL Each for $61.13
03-700C
  Accessories
fisherbrand™
703700C
1.69 oz. ±0.1mL 50mL 0.1mL Each for $14.90
Case of 12 Each for $178.80
03-700D
  Accessories
fisherbrand™
703700D
3.38 oz. ±0.2mL 100mL 0.2mL Each for $187.80
Description & Specifications
Specifications
Product Type Manual Buret
For Use With (Application) For both educational and routine laboratory work
Includes Straight stopcock, blue markings
Stopcock Material Glass
  • Well suited for both educational and routine laboratory work