missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Magnetic Stir Bar Six Packs

These magnetic stirring bars with integral pivot ring are the most commonly used shape.

Manufacturer:  Fisherbrand™ 800371117

 View more versions of this product

Catalog No. 1451351SIX



Description

Description

These magnetic stirring bars with integral pivot ring are the most commonly used shape.

 

  • Alnico magnet encapsulated in FDA Grade, USP Class VI Teflon™ PTFE low friction coating; durable and inert
  • Interrupted profile provides greater surface area and added turbulence when compared to the smooth surface of cylindrical bars
  • Pivot ring aids in reducing friction and chattering
  • Autoclavable
  • Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification

 

Specifications

Specifications

Yes
White
1 in.
9.5mm
Alnico Magnet
PTFE
0.375 in.
Stir bar
25.4mm
Documents