Please login to your online account to display your discounted pricing
Fisherbrand™ Magnetic Micro Stirring Bars
Micro magnetic stirring bars are designed for stirring small volumes in vessels such as vials, tubes and gradient makers
$2.50 - $3.90
Specifications
Coating PTFE
Material Alnico Magnet
Product Type Micro Stir Bar
View More Specs
Products
Catalog Number Mfr. No. Diameter (English) Diameter (Metric) Length (English) Length (Metric) Price Quantity    
14-513-63
  Accessories
fisherbrand™
371195001
0.07 in. 2mm 0.27 in. 7mm Each for $3.90
Case of 50 Each for $195.00
14-513-65
  Accessories
fisherbrand™
371195003
0.11 in. 3mm 0.39 in. 10mm Each for $2.70
Case of 50 Each for $135.00
14-513-98
  Accessories
fisherbrand™
371105016
0.11 in. 3mm 0.5 in. 12.7mm Each for $2.50
Case of 50 Each for $125.00
14-513-64
  Accessories
fisherbrand™
371195002
0.06 in. 1.5mm 0.31 in. 8mm Each for $3.90
Case of 50 Each for $195.00
Description & Specifications
Specifications
Coating PTFE
Material Alnico Magnet
Product Type Micro Stir Bar
  • Particularly useful for environmental testing and life science applications in which small sample volumes need to be prepared and evaluated
  • Alnico magnet encapsulated in FDA Grade, USP Class VI Teflon® PTFE low friction coating; durable and inert
  • Manufactured in a registered ISO 9001:2008, as verified by SGS certification
  • Autoclave safe
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.