Please login to your online account to display your discounted pricing

Fisherbrand™ Machlett Automatic Burets


Class A, Gravity filling eliminates errors and guards against deterioration of liquids affected by air

Manufacturer: fisher scientific  TG1831101

 View more versions of this product

Catalog No. 03-847-1A

  • / Each
 

Description & Specifications

Specifications

Subdivision 0.01mL
Tolerance ±0.01mL
Capacity (Metric) 2mL
Product Type Machlett Automatic Buret
For Use With (Application) Used in multiples by connecting central storage vessel to each reservoir neck opening when used in this way, detachable drying tube is placed on storage vessel
Includes PTFE plug stopcock
Material Glass

  • Half turn of PTFE-plug stopcock fills buret; another half turn dispenses liquid
  • Overflow is automatically drawn back into reservoir through tube connecting reservoir with overflow tube
  • With stopcock handle in horizontal position, all action stops
  • Single air intake on reservoir neck is protected by drying tube, which removes moisture from incoming air
  • Can be used in multiples by connecting central storage vessel to each reservoir neck opening—when used in this way, detachable drying tube is placed on storage vessel