Please login to your online account to display your discounted pricing
Fisherbrand™ Machlett Automatic Burets
Class A, Gravity filling eliminates errors and guards against deterioration of liquids affected by air
$1,341.67 - $1,863.33
Specifications
Product Type Machlett Automatic Buret
For Use With (Application) Used in multiples by connecting central storage vessel to each reservoir neck opening when used in this way, detachable drying tube is placed on storage vessel
Includes PTFE plug stopcock
Material Glass
View More Specs
Products
Catalog Number Mfr. No. Capacity (Metric) Subdivision Tolerance Price Quantity    
03-847-1A
fisher scientific
TG1831101
2mL 0.01mL ±0.01mL Each for $1,341.67
03-847-1B
fisher scientific
TG1831102
5mL 0.02mL ±0.01mL Each for $1,547.63
03-847-1C
fisher scientific
TG1831103
10mL 0.05mL ±0.02mL Each for $1,544.57
03-847-1D
fisher scientific
TG1831104
25mL 0.10mL ±0.03mL Each for $1,783.27
03-847-1E
fisher scientific
TG1831105
50mL 0.10mL ±0.05mL Each for $1,863.33
Description & Specifications
Specifications
Product Type Machlett Automatic Buret
For Use With (Application) Used in multiples by connecting central storage vessel to each reservoir neck opening when used in this way, detachable drying tube is placed on storage vessel
Includes PTFE plug stopcock
Material Glass
  • Half turn of PTFE-plug stopcock fills buret; another half turn dispenses liquid
  • Overflow is automatically drawn back into reservoir through tube connecting reservoir with overflow tube
  • With stopcock handle in horizontal position, all action stops
  • Single air intake on reservoir neck is protected by drying tube, which removes moisture from incoming air
  • Can be used in multiples by connecting central storage vessel to each reservoir neck opening—when used in this way, detachable drying tube is placed on storage vessel