Please login to your online account to display your discounted pricing

Fisherbrand™ Low-Retention Pipette Tips, Nonfilter


Increase accuracy by reducing sample retention

Manufacturer: fisherbrand™  110BNLRFIS

 View more versions of this product

Catalog No. 02-717-139

  • / Pack of 1000 Each
  • / Case of 10 Pack
 

Description & Specifications

Specifications

Sterility Nonsterile
For Use With (Equipment) Universal Fit
Packaging Bulk Pack
Volume (Metric) 200μL
Color Natural
Model Low retention
Tip Style Beveled