missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Low-Retention Pipette Tips, Nonfilter

Increase accuracy by reducing sample retention.

$118.25 - $166.07

Specifications

Color Natural
For Use With (Equipment) Universal Fit
Material Polypropylene
View More Specs

Products 3
Catalog Number Mfr. No. Volume (Metric) Sterility Price Quantity  
Catalog Number Mfr. No. Volume (Metric) Sterility Price Quantity  
02-717-133
View Documents
N/A 10 μL Non-sterile
Pack of 1000 Each for $166.07
Case of 10 Pack for $1,660.70
Only null left
 
02-717-144
View Documents
N/A 200 μL Sterile
Pack of 960 Each for $118.25
Case of 5 Pack for $591.25
Only null left
 
02-717-134
View Documents
N/A 0.1 to 10 μL Non-sterile
Pack of 960 Each for $149.79
Case of 5 Pack for $748.95
Only null left
 
Description

Description

Increase accuracy by reducing sample retention

  • Low retention surface minimizes samples loss
  • Certified by independent laboratory
  • Nonsterile: RNase, DNase, DNA, and Pyrogen Free; Sterile: RNase, DNase, DNA, Bioburden, PCR Inhibitor, and Pyrogen Free
  • Natural color
Specifications

Specifications

Natural
Universal Fit
Polypropylene
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'